Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002019

Ropinirole for peak identification 3

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ropinirole hydrochloride, 4-[2-(Dipropylamino)ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride, SKF 101468 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$169.00

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$169.00

About This Item

Empirical Formula (Hill Notation):
C16H24N2O · HCl
CAS Number:
Molecular Weight:
296.84
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$169.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ropinirole

manufacturer/tradename

EDQM

format

neat

storage temp.

2-8°C

SMILES string

Cl.CCCN(CCC)CCc1cccc2NC(=O)Cc12

InChI

1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H

InChI key

XDXHAEQXIBQUEZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ropinirole for peak identification 3 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

An agonist at the D2 and D3 dopamine receptor subtypes, binding with higher affinity to D3 than to D2 or D4. It has negligible effect on D1-receptors. It has medium in vitro affinity to opioid receptors. Ropinirole is said to have virtually no affinity to 5-HT1, 5-HT2, benzodiazepine, GABA, muscarinic, α1-, α2-, and β-adrenoreceptors. Used as antiparkinsonian drug.
Selective D2 dopamine receptor agonist, antiparkinsonian

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service