Y0001870
Andrographis herb HRS
European Pharmacopoeia (EP) Reference Standard
Select a Size
$335.00
Select a Size
About This Item
$335.00
Recommended Products
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Packaging
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service