Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001839

Hydroxychloroquine sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-Chloro-4-[4-(N-ethyl-N-b-hydroxyethylamino)-1-methylbutylamino]quinoline sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

70 MG
$335.00

$335.00


Please contact Customer Service for Availability


Select a Size

Change View
70 MG
$335.00

About This Item

Empirical Formula (Hill Notation):
C18H26ClN3O · H2SO4
CAS Number:
Molecular Weight:
433.95
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$335.00


Please contact Customer Service for Availability

API family

hydroxychloroquine

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12

InChI

1S/C18H26ClN3O.H2O4S/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;1-5(2,3)4/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);(H2,1,2,3,4)

InChI key

JCBIVZZPXRZKTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Hydroxychloroquine sulfate is an anti-inflammatory (via TNF), disease-modifying antirheumatic drug (DMARD), and a heme polymerase inhibitor.
Hydroxychloroquine sulfate is an anti-inflammatory (via TNF), disease-modifying antirheumatic drug (DMARD), and a heme polymerase inhibitor. It is used in treating malaria.

Packaging

Unit quantity: 70 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service