Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001791

Thiocolchicoside for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Thiocolchicoside, (S)-N-[3-(β-D-glucopyranosyloxy)-1,2-dimethoxy-10(methylthio)-9-oxo-5,6,7,9-tetrahydrobenzo [a] heptalen-7-yl] acetamide, 2-Demethoxy-2-glucosidoxythiocolchicine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C27H33NO10S
CAS Number:
Molecular Weight:
563.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

thiocolchicoside

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1[C@H](O)[C@@H](CO)O[C@@H](OC2=CC(CC[C@H](NC(C)=O)C(C3=CC=C4SC)=CC4=O)=C3C(OC)=C2OC)[C@@H]1O

InChI

1S/C27H33NO10S/c1-12(30)28-16-7-5-13-9-18(37-27-24(34)23(33)22(32)19(11-29)38-27)25(35-2)26(36-3)21(13)14-6-8-20(39-4)17(31)10-15(14)16/h6,8-10,16,19,22-24,27,29,32-34H,5,7,11H2,1-4H3,(H,28,30)/t16-,19+,22+,23-,24+,27+/m0/s1

InChI key

LEQAKWQJCITZNK-AXHKHJLKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thiocolchicoside for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Thiocolchicoside is a potent competitive γ-aminobutyric acid type A (GABAA) receptor antagonist and glycine receptor agonist with muscle relaxant, anti-inflammatory, and analgesic activity.
Thiocolchicoside is a potent competitive γ-aminobutyric acid type A (GABAA) receptor antagonist and glycine receptor agonist with muscle relaxant, anti-inflammatory, and analgesic activity. Thiocolchicoside has also been found to suppress nuclear factor kappa B ligand (RANKL) activation of NF-KB resulting in suppression of osteoclastogenesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Target Organs

Gastro-intestinal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service