Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001772

Sodium lauroylsarcosinate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Lauroylsarcosine sodium salt, N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing

Select a Size

550 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
550 MG
$205.00

About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein:
5322974
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sarcosinates

mol wt

micellar avg mol wt 600

manufacturer/tradename

EDQM

aggregation number

2

CMC

14.6 mM (20-25°C)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium lauroylsarcosinate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Anionic surfactant.
Sales restrictions may apply.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

512.6 °F - closed cup

Flash Point(C)

267 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How should I store the solution once I have resuspended the powder?

    1 answer
    1. This product is a Reference Standard supplied by the European Pharmacopoeia (EP) and is stored at refrigerated at 2-8°C. Solution storage conditions are not provided. This compound is also available as a prepared 20% solution for molecular biology use under product L7414. This research grade product may be stored at room temperature for at least 1 year. Similar stability would be expected for this item. Please see the link below to review the product details:
      https://www.sigmaaldrich.com/product/sigma/l7414

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service