Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001572

Atovaquone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Mepron, trans-2-[4-(4-Chlorophenyl)cyclohexyl]-3-hydroxy-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing

Select a Size

270 MG
$204.00

$204.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
270 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C22H19ClO3
CAS Number:
Molecular Weight:
366.84
UNSPSC Code:
41116107
NACRES:
NA.24

$204.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

atovaquone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC1=C([C@H]2CC[C@@H](CC2)c3ccc(Cl)cc3)C(=O)c4ccccc4C1=O

InChI

1S/C22H19ClO3/c23-16-11-9-14(10-12-16)13-5-7-15(8-6-13)19-20(24)17-3-1-2-4-18(17)21(25)22(19)26/h1-4,9-13,15,26H,5-8H2/t13-,15-

InChI key

KUCQYCKVKVOKAY-CTYIDZIISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Atovaquone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Atovaquone in an anti-protozoal mitochondrial electron transport inhibitor. It also functions as an antimalarial and antipneumocystic agent.
Atovaquone is an anti-protozoal mitochondrial electron transport inhibitor; Antimalarial; Antipneumocystic, and has also been used to treat toxoplasmosis. It is an analog of protozoan mitochondrial protein ubiquinone, and acts by inhibiting the cytochrome bc(1) complex via interactions with the Rieske iron-sulfur protein and cytochrome b in the ubiquinol oxidation pocket.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shirly Grynberg et al.
The American journal of tropical medicine and hygiene, 92(1), 13-17 (2014-11-06)
Atovaquone-proguanil (AP) and artemether-lumefantrine (AL) are both treatments for uncomplicated Plasmodium falciparum malaria, but comparative clinical trials are lacking. We performed a retrospective analysis, comparing treatment failure and fever clearance time in non-immune travelers with uncomplicated P. falciparum malaria, treated
Geoffrey W Birrell et al.
Antimicrobial agents and chemotherapy, 59(1), 170-177 (2014-10-22)
4-(tert-Butyl)-2-((tert-butylamino)methyl)-6-(6-(trifluoromethyl)pyridin-3-yl)-phenol (JPC-2997) is a new aminomethylphenol compound that is highly active in vitro against the chloroquine-sensitive D6, the chloroquine-resistant W2, and the multidrug-resistant TM90-C2B Plasmodium falciparum lines, with 50% inhibitory concentrations (IC50s) ranging from 7 nM to 34 nM. JPC-2997
L M Upton et al.
Antimicrobial agents and chemotherapy, 59(1), 490-497 (2014-11-12)
To achieve malarial elimination, we must employ interventions that reduce the exposure of human populations to infectious mosquitoes. To this end, numerous antimalarial drugs are under assessment in a variety of transmission-blocking assays which fail to measure the single crucial
Sanna R Rijpma et al.
Malaria journal, 13, 359-359 (2014-09-15)
Therapeutic blood plasma concentrations of anti-malarial drugs are essential for successful treatment. Pharmacokinetics of pharmaceutical compounds are dependent of adsorption, distribution, metabolism, and excretion. ATP binding cassette (ABC) transport proteins are particularly involved in drug deposition, as they are located
F Giallongo et al.
Journal of dairy science, 98(5), 3292-3308 (2015-03-03)
This experiment was conducted with the objective to investigate the effects of slow-release urea and rumen-protected (RP) Met and His supplementation of a metabolizable protein (MP)-deficient diet (according to NRC, 2001) on lactation performance of dairy cows. Sixty lactating Holstein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service