Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001376

2-Methylpyrazine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105778
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

glucosamine

manufacturer/tradename

EDQM

refractive index

n20/D 1.504 (lit.)

bp

135 °C/761 mmHg (lit.)

mp

−29 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cc1cnccn1

InChI

1S/C5H6N2/c1-5-4-6-2-3-7-5/h2-4H,1H3

InChI key

CAWHJQAVHZEVTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

2-Methylpyrazine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruna R Toledo et al.
Journal of chromatography. A, 1346, 1-7 (2014-05-03)
Solid-phase microextraction (SPME) using cross-linked polymeric ionic liquid (PIL)-based sorbent coatings was used to extract volatile aroma-related compounds from coffee samples. Several PIL-based coatings were screened alongside a commercial poly(acrylate) (PA) SPME coating. The best performing PIL-based SPME fiber, poly(1-vinyl-3-hexadecylimidazolium

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service