Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001171

Astragaloside IV

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

40 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C41H68O14
CAS Number:
Molecular Weight:
784.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

astragaloside

manufacturer/tradename

EDQM

mp

295-296 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(O)[C@@H]1CC[C@@](C)(O1)[C@H]2[C@@H](O)C[C@@]3(C)[C@@H]4C[C@H](O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@H]6C(C)(C)[C@H](CC[C@@]67C[C@@]47CC[C@]23C)O[C@@H]8OC[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C41H68O14/c1-35(2)24(54-33-29(48)26(45)20(44)17-51-33)9-11-41-18-40(41)13-12-37(5)31(39(7)10-8-25(55-39)36(3,4)50)19(43)15-38(37,6)23(40)14-21(32(35)41)52-34-30(49)28(47)27(46)22(16-42)53-34/h19-34,42-50H,8-18H2,1-7H3/t19-,20+,21-,22+,23-,24-,25-,26-,27+,28-,29+,30+,31-,32-,33-,34+,37+,38-,39+,40-,41+/m0/s1

InChI key

QMNWISYXSJWHRY-YLNUDOOFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Astragaloside IV EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anti-inflammatory.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Liu et al.
Archivum immunologiae et therapiae experimentalis, 68(6), 33-33 (2020-10-24)
Astragaloside IV (AS-IV) has shown anti-tumorigenic properties in certain cancers for its effect of boosting the body's immune system, but its role in colorectal cancer (CRC) remains unclear. In this study, we investigated the therapeutic effect of AS-IV in CRC
Shuang Ren et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 33(3), 413-416 (2013-09-13)
To review the pharmacological effects and mechanisms of action of Astragaloside IV in Huangqi (Radix Astragali Mongolici). Aticles focusing on Astragaloside IV in English and Chinese in databases were collected and reviewed in order to summarize the latest extraction separation
Jing Zhang et al.
FEMS microbiology letters, 367(4) (2020-02-29)
Excessive secretion of inflammatory factors (cytokine storm) plays a significant role in H1N1-induced acute pneumonia, and autophagy acts as a cell-intrinsic mechanism to regulate inflammation. Astragaloside IV (AS-IV), originating from the astragalus root, possesses multiple pharmacological activities, such as anti-inflammation.
Biwang Liu et al.
Pharmacology, 105(1-2), 90-101 (2019-09-26)
Astragaloside IV has shown its promising effect on acute respiratory distress syndrome (ARDS). We aim to explore whether astragaloside IV is effective for ARDS treatment in a lipopolysaccharides (LPS)-induced cell model and whether autophagy is involved in the therapeutic function
Li Wang et al.
Experimental biology and medicine (Maywood, N.J.), 239(10), 1310-1324 (2014-06-01)
Astragaloside IV (AS-IV) is a major active ingredient from Radix astragali, which has been considered as a renoprotective agent; however, its molecular mechanisms are unclear. Thus, we designed to investigate the renoprotective effects and mechanisms of AS-IV in rat model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service