Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001127

Phentolamine mesilate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Phentolamine methanesulfonate salt, 2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
377.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phentolamine

manufacturer/tradename

EDQM

mp

180-182 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3

InChI

1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)

InChI key

OGIYDFVHFQEFKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phentolamine mesilate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

α-Adrenergic receptor antagonist; peripheral vasodilator.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianming Su et al.
Journal of cardiovascular pharmacology, 65(1), 62-71 (2014-09-30)
Previous studies have found that Danhong injection (DHI), an extensively used herbal extract preparation in China, might be a powerful vasodilator. The aims of this study were to determine the vascular activity of DHI and its effects on arteries of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service