Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001055

Tributyl acetylcitrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Tributyl 2-acetylcitrate, Acetyl tributyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
Beilstein:
2303316
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

0.26 psi ( 20 °C)
0.8 psi ( 55 °C)

description

API family: acetyltributyl citrate

API family

acetyltributyl citrate

manufacturer/tradename

EDQM

refractive index

n20/D 1.443 (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC

InChI

1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3

InChI key

QZCLKYGREBVARF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Tributyl acetylcitrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

422.6 °F - closed cup

Flash Point(C)

217 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service