Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000618

Tropisetron impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Indole-3-carboxylic acid, β-Indolylcarboxylic acid, 3-Carboxyindole, 3-Indole formic acid, 3-Indolylcarboxylic acid, Indole-β-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$299.00

$299.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$299.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
Beilstein:
129435
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$299.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

tropisetron

manufacturer/tradename

EDQM

mp

232-234 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)c1c[nH]c2ccccc12

InChI

1S/C9H7NO2/c11-9(12)7-5-10-8-4-2-1-3-6(7)8/h1-5,10H,(H,11,12)

InChI key

KMAKOBLIOCQGJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Reactant for preparation of:
  • Anticancer agents
  • Derivatives of amino acids and peptides
  • Serotonin 5-HT4 receptor antagonists
  • Primary acylureas
  • Inhibitors of Gli1-mediated transcription in the Hedgehog pathway
  • Serotonin 5-HT6 antagonists
  • Very Late Antigen-4 (VLA-4) sntagonists
  • EphB3 receptor tyrosine kinase inhibitors
  • Potential Therapeutic Agent for Alzheimer′s Disease
  • Vinyl ester pseudotripeptide proteasome inhibitors

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service