Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000092

Trimetazidine dihydrochloride - reference spectrum

European Pharmacopoeia (EP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

reference spectrum

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: Reference spectrum

Other Notes

This product is not a physical sample. It is a document containing spectrographic information.
Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiwen Lu et al.
Journal of receptor and signal transduction research, 41(2), 170-179 (2020-08-08)
Previous studies demonstrated the effect of Trimetazidine (TMZ) on alleviating cardiomyocytes Hypoxia/Reoxygenation (H/R) injuries and the protective effect of autophagy on Ischemia-Reperfusion (I/R) cell injuries. However, whether the protection mechanism of TMZ was also involved in autophagy remained unclear. Our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service