Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W379620

Sigma-Aldrich

Vanillyl butyl ether

≥96%, FG

Synonym(s):

4-(butoxymethyl)-2-methoxy-Phenol

Sign Into View Organizational & Contract Pricing

Select a Size

W379620-SAMPLE
$74.30
100 G
$146.00
1 KG
$837.00
5 KG
$3,640.00

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W379620-SAMPLE
$74.30
100 G
$146.00
1 KG
$837.00
5 KG
$3,640.00

About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
210.27
FEMA Number:
3796
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.093
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
biological source:
synthetic
food allergen:
no known allergens

$74.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥96%

refractive index

n20/D 1.516 (lit.)

bp

241 °C (lit.)

density

1.057 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

creamy; vanilla

SMILES string

CCCCOCC1=CC(OC)=C(O)C=C1

InChI

1S/C12H18O3/c1-3-4-7-15-9-10-5-6-11(13)12(8-10)14-2/h5-6,8,13H,3-4,7,9H2,1-2H3

InChI key

VLDFMKOUUQYFGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Inducible desensitization to capsaicin with repeated low-dose exposure in human volunteers.: This randomized controlled trial explores the desensitization effects of capsaicin through repeated low-dose exposure. The study provides insights into the potential applications of vanillyl butyl ether as an alternative for reducing sensitivity to capsaicin. (Nolden et al., 2024).

  • The effects of vanilloid analogues structurally related to capsaicin on the transient receptor potential vanilloid 1 channel.: This research investigates the impact of various vanilloid analogues, including vanillyl butyl ether, on the TRPV1 channel, highlighting its biochemical interactions and potential therapeutic applications. (Oka et al., 2022).

  • Vertical alignment of liquid crystals on plant-based vanillin derivative-substituted polystyrene films.: This study explores the use of vanillin derivatives, including vanillyl butyl ether, in the alignment of liquid crystals, which could lead to advancements in display technologies. (Ju et al., 2019).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service