Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W273601

Sigma-Aldrich

Phenyl ethyl dimethyl carbinyl isobutyrate

≥97%, FG

Synonym(s):

papaya isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O2
CAS Number:
Molecular Weight:
234.33
FEMA Number:
2736
EC Number:
Council of Europe no.:
2086
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.484
NACRES:
NA.21
Organoleptic:
floral; fruity; sweet
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

floral; fruity; sweet

SMILES string

CC(C)(OC(C(C)C)=O)CCC1=CC=CC=C1

InChI

1S/C15H22O2/c1-12(2)14(16)17-15(3,4)11-10-13-8-6-5-7-9-13/h5-9,12H,10-11H2,1-4H3

InChI key

WCEXWNUHYPYHDN-UHFFFAOYSA-N

Related Categories

General description

Phenyl ethyl dimethyl carbinyl isobutyrate is an aryl alkyl alcohol simple acid ester. It is also referred to as 2-methyl-4-phenyl-2-butyl isobutyrate. Vapor pressure of phenyl ethyl dimethyl carbinyl isobutyrate is 0.00187mmHg at 25°C. Safety and dermatological assessment of phenyl ethyl dimethyl carbinyl isobutyrate have been reported.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Belsito et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S269-S313 (2012-03-13)
The aryl alkyl alcohol simple acid ester derivatives (AAASAE) group of fragrance ingredients was critically evaluated for safety following a complete literature search of the pertinent data. For high end users, calculated maximum skin exposures vary widely from 0.01% to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service