Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VISCWAT

Pure Water Viscosity Standard

certified at 5 °C, 20 °C, 25 °C & 37 °C. UKAS ISO 17025 and ISO 17034 certified

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 100 mL

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation

Paragon ISO 17025 / ISO 17034 certified pure water viscosity standards are specifically designed, tested and certified for the calibration, verification and certification of glass capillary viscometers and other viscosity measuring equipment where the precise control of temperature is required.

ASTM D1480 / ASTM D2162

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service