Key Documents
UBBEL04NC
Ubbelohde Viscometer for Transparent Liquids
Size 1, Nom. Constant 0.01, Range 2 to 10 cSt, (non certified)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
General description
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product UBBEL04UKC
Application
Ubbelohde Viscometer may be used for the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D 445, ISO 3104, IP 71, and BS188.
Choose from one of the most recent versions:
Certificates of Analysis (COA)