Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7645

Sigma-Aldrich

Tetracaine hydrochloride

Sigma Reference Standard

Synonym(s):

4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester, Amethocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2 · HCl
CAS Number:
Molecular Weight:
300.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Sigma Reference Standard

packaging

vial of 250 mg

solubility

alcohol: soluble in 40 parts solvent
chloroform: soluble in 30 parts of solvent
H2O: soluble 50 mg/mL, clear, colorless
acetone: insoluble
benzene: insoluble
diethyl ether: insoluble

SMILES string

Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C

InChI

1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H

InChI key

PPWHTZKZQNXVAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetracaine hydrochloride has been used to analyze Ca2+ release from sarcoplasmic reticulum (SR) in neonatal ventricular cardiomyocytes (NVCMs). Tetracaine hydrochloride has also been used as a component of protease digestion buffers.
Topical ophthalmic anesthetic; used for spinal anesthesia

Biochem/physiol Actions

Blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum.

Packaging

Supplied in amber screw-cap vials

Preparation Note

Tetracaine hydrochloride dissolves in water at 50 mg/ml to yield a clear, colorless solution. It is also soluble in 40 parts alcohol and 30 parts chloroform. However, it is practically insoluble in ether benzene or acetone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Knight et al.
European journal of immunology, 22(3), 879-882 (1992-03-01)
Recent reports have shown that both exogenous and endogenous mouse mammary tumor viruses (MMTV) can encode superantigens. Transfection and transgenic studies have identified the open reading frame (ORF) present in the 3' long terminal repeat (LTR) as encoding superantigen function.
Patrick Most et al.
Journal of cell science, 118(Pt 2), 421-431 (2005-01-18)
Calcium is a key regulator of cardiac function and is modulated through the Ca2+-sensor protein S100A1. S100 proteins are considered to exert both intracellular and extracellular functions on their target cells. Here we report the impact of an increased intracellular
Patrick E McCleskey et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 39(1 Pt 1), 82-91 (2013-01-03)
Few published studies have analyzed serum lidocaine levels and individual patient characteristics affecting metabolism after application of compounded topical anesthetics. To measure serum lidocaine levels during and cutaneous side effects after standardized application of 23% lidocaine/7% tetracaine compounded anesthetic to
Gabriel C Gaviola et al.
The Laryngoscope, 123(4), 852-858 (2013-02-14)
Transnasal endoscopy is commonly performed in an outpatient otolaryngology setting. Patients are typically administered a topical anesthetic and decongestant prior to this procedure to alleviate discomfort and improve visualization. There is no consensus on which topical anesthetic is most effective
K Ruetzler et al.
British journal of anaesthesia, 109(5), 790-796 (2012-07-27)
Arterial catheterization is painful and is associated with patient stress and anxiety. Analgesia is usually provided by subcutaneous injection of local anaesthetic. An alternative is topical anaesthesia, such as Rapydan which is a novel topical anaesthetic patch containing 70 mg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service