Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB01348

Sigma-Aldrich

Dihydrocortisol

≥95% (HPLC)

Synonym(s):

5β-Pregnane-11β,17α,21-triol-3,20-dione, 11β,17,21-Trihydroxy-5β-pregnane-3,20-dione, 11β,17α,21-Trihydroxy-5β-pregnane-3,20-dione, 5β-Dihydrocortisol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$152.00
100 MG
$338.00

$152.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$152.00
100 MG
$338.00

About This Item

Empirical Formula (Hill Notation):
C21H32O5
CAS Number:
Molecular Weight:
364.48
MDL number:
UNSPSC Code:
51420000

$152.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

O[C@]1([C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](CC3)CC(=O)CC4)C)[C@H](C2)O)CC1)C)C(=O)CO

InChI

1S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12,14-16,18,22,24,26H,3-11H2,1-2H3/t12-,14+,15+,16+,18-,19+,20+,21+/m1/s1

InChI key

ACSFOIGNUQUIGE-AIPUTVCKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5β-Dihydrocortisol is the product of the enzyme steroid 5-beta-reductase, which catalyzes the reduction of progesterone, androstenedione, 17-alpha-hydroxyprogesterone, testosterone, aldosterone, corticosterone, and cortisol to 5-beta-reduced metabolites. 5β-Dihydrocortisol has shown to potentiate the action of topically applied dexamethasone in raising the intraocular pressure in young rabbits.

Application

Metabolomics research

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Southren et al.
Investigative ophthalmology & visual science, 26(3), 393-395 (1985-03-01)
5 beta-dihydrocortisol potentiates the action of topically applied dexamethasone in raising the intraocular pressure (IOP) in young rabbits. Dexamethasone (0.06%) plus 5 beta-dihydrocortisol (0.1 and 1.0%) elevated the IOP 7-10 mmHg within 18 days of treatment. By contrast, 0.06% dexamethasone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service