Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SMB00969

Sigma-Aldrich

(E)-10-Hydroxy amitriptyline

≥95% (HPLC)

Synonym(s):

Amitriptyline metabolite, (±)-E-10-hydroxylated-, Amitriptyline, (±)-(E)-5-[3-(Dimethylamino)propylidene]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$805.00

$805.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$805.00

About This Item

Empirical Formula (Hill Notation):
C20H23NO
CAS Number:
Molecular Weight:
293.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

$805.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

technique(s)

HPLC: suitable

color

white to yellow

storage temp.

2-8°C

SMILES string

CN(C)CC\C=C1/c2ccccc2CC(O)c3ccccc13

InChI

1S/C20H23NO/c1-21(2)13-7-12-17-16-9-4-3-8-15(16)14-20(22)19-11-6-5-10-18(17)19/h3-6,8-12,20,22H,7,13-14H2,1-2H3/b17-12+

InChI key

GHWBJXOKAFHZAI-SFQUDFHCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(E)-10-Hydroxy amitriptyline is a metabolite of Amitriptyline (brand name Elavil), a FDA-approved oral medicine for the treatment of major depressive and anxiety disorders, attention deficit hyperactivity disorder (ADHD), bipolar disorder, as well as migraines and neuropathic pain (fibromyalgia & postherpetic neuralgiapain). Amitriptyline acts mainly on the serotonin transporter (SERT) and moderately on the norepinephrine transporter (NET). Amitriptyline is metabolised by cytochrome P450 enzymes (demethylation by CYP2C19 & CYP3A4) in the liver to Nortriptyline, which also acts as a noradrenaline reuptake inhibitor and thereby potentiates the antidepressant effects of amitriptyline. Amitriptyline and Nortriptyline are subject to metabolisation by hydroxylation (by CYP2D6) and n-oxidation.

Application

Metabolomics research

Biochem/physiol Actions

Metabolite of amitriptyline, tricyclic antidepressant; chromatographic standard.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ursula Breyer-Pfaff
Drug metabolism reviews, 36(3-4), 723-746 (2004-11-24)
Amitriptyline (AT), the most widely used tricyclic antidepressant, undergoes oxidative metabolism in the side chain with production of the secondary amine nortriptyline (NT), a primary amine, and the N-oxide amitriptylinoxide (AT-NO); in addition, direct conjugation leads to a quaternary ammonium-linked

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service