Skip to Content
MilliporeSigma
All Photos(1)

Documents

S5020

Sigma-Aldrich

Silver enhancer solution A

liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

product name

Silver enhancer solution A,

form

liquid

Quality Level

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

Application

Silver enhancer solution A has been used for silver staining.

Other Notes

Silver salt solution for Silver Enhancer Kit SE-100

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly sensitive detection of cardiac troponin I in human serum using gold nanoparticle-based enhanced sandwich immunoassay
Wan-Joong Kim
Sensors & Transducers Journal, 221 (2015)
Multiplexed electrochemical immunoassay using streptavidin/nanogold/carbon nanohorn as a signal tag to induce silver deposition
Changrong Zhao
Analytica Chimica Acta, 37-43 (2014)
Jeong Moon et al.
Macromolecular bioscience, 19(6), e1800486-e1800486 (2019-04-19)
For the construction of high-performance biosensor, it is important to interface bioreceptors with the sensor surface densely and in the optimal orientation. Herein, a simple surface modification method that can optimally immobilize antibodies onto various kinds of surfaces is reported.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service