Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S3259

Sigma-Aldrich

Solvent Blue 37

practical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H16N3Na3O11S3
CAS Number:
Molecular Weight:
711.58
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

practical grade

SMILES string

Oc1cc(Nc2ccccc2)c3c(cccc3c1N=Nc4cc(cc5cc(cc(O)c45)S(=O)(=O)O[Na])S(=O)(=O)O[Na])S(=O)(=O)O[Na]

InChI

1S/C26H19N3O11S3.3Na/c30-21-12-17(42(35,36)37)10-14-9-16(41(32,33)34)11-20(24(14)21)28-29-26-18-7-4-8-23(43(38,39)40)25(18)19(13-22(26)31)27-15-5-2-1-3-6-15;;;/h1-13,27,30-31H,(H,32,33,34)(H,35,36,37)(H,38,39,40);;;/q;3*+1/p-3

InChI key

LONJZSVIJVYVID-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service