Skip to Content
MilliporeSigma
All Photos(1)

Documents

S2159100

Sulfapyridine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Amino-N-[2-pyridyl]benzene sulfonamide, N1-(Pyridin-2-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3O2S
CAS Number:
Molecular Weight:
249.29
Beilstein:
222065
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfapyridine

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2

InChI

1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)

InChI key

GECHUMIMRBOMGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfapyridine is a metabolite of Sulfasalazine. It is an antibiotic and is mostly used in treating rheumatoid arthritis.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfapyridine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dapsone and sulfapyridine.
J Uetrecht
Clinics in dermatology, 7(3), 111-120 (1989-07-01)
Treatment with sulfasalazine or sulfapyridine, but not 5-aminosalicylic acid, inhibits basic fibroblast growth factor-induced endothelial cell chemotaxis.
Volin, Michael V., et al.
Arthritis and Rheumatism, 42.9, 1927-1935 (1999)
U Paniker et al.
Dermatologic clinics, 19(1), 79-86 (2001-01-13)
Dapsone and sulfapyridine are structurally related compounds with anti-microbial and anti-inflammatory effects. Dapsone remains the most important drug for leprosy and is useful in the prophylaxis of Pneumocystis pneumonia in patients with HIV disease. The medical treatment of choice for
Shaojun Yang et al.
The Journal of pharmacology and experimental therapeutics, 352(3), 529-540 (2015-01-01)
Sepiapterin reductase (SPR) catalyzes the reduction of sepiapterin to dihydrobiopterin (BH2), the precursor for tetrahydrobiopterin (BH4), a cofactor critical for nitric oxide biosynthesis and alkylglycerol and aromatic amino acid metabolism. SPR also mediates chemical redox cycling, catalyzing one-electron reduction of
M Díaz-Álvarez et al.
Journal of chromatography. A, 1357, 158-164 (2014-05-02)
In this work, molecularly imprinted polymer (MIP) beads have been prepared and evaluated for the development of a supported liquid membrane-protected micro-solid phase extraction method for the analysis of sulfonamides (SAs) in aqueous samples. The performance of MIP beads was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service