Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S1700000

Sulfacetamide sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sulfacetamide sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$205.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sulfacetamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na]N([S](=O)(=O)c1ccc(cc1)N)C(=O)C

InChI

1S/C8H10N2O3S.Na/c1-6(11)10-14(12,13)8-4-2-7(9)3-5-8;/h2-5H,9H2,1H3,(H,10,11);/q;+1/p-1

InChI key

PQMSFAORUFMASU-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfacetamide sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of alizarine derivatives as chromogenic reagents for the spectrophotometric determination of some sulfa drugs.
Amin S A, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 51(3), 367-373 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service