Skip to Content
MilliporeSigma
All Photos(1)

Documents

S1148

Sigma-Aldrich

SBFI-AM

≥80% purity

Synonym(s):

Sodium indicator SBFI-AM, Sodium-binding benzofuran isophthalate-AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H58N2O23
CAS Number:
Molecular Weight:
1127.06
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

SBFI-AM,

storage temp.

−20°C

Quality Level

SMILES string

COc1cc2cc(oc2cc1N3CCOCCOCCN(CCOCC3)c4cc5oc(cc5cc4OC)-c6ccc(cc6C(=O)OCOC(C)=O)C(=O)OCOC(C)=O)-c7ccc(cc7C(=O)OCOC(C)=O)C(=O)OCOC(C)=O

InChI

1S/C56H58N2O23/c1-33(59)72-29-76-53(63)37-7-9-41(43(21-37)55(65)78-31-74-35(3)61)49-23-39-25-51(67-5)45(27-47(39)80-49)57-11-15-69-16-12-58(14-18-71-20-19-70-17-13-57)46-28-48-40(26-52(46)68-6)24-50(81-48)42-10-8-38(54(64)77-30-73-34(2)60)22-44(42)56(66)79-32-75-36(4)62/h7-10,21-28H,11-20,29-32H2,1-6H3

InChI key

MWUYLQGOGOLRQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cell permeant sodium selective fluorescent indicator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Cristina Brailoiu et al.
Drug and alcohol dependence, 178, 7-14 (2017-06-18)
HIV-1 infection and drug abuse are frequently co-morbid and their association greatly increases the severity of HIV-1-induced neuropathology. While nucleus accumbens (NAcc) function is severely perturbed by drugs of abuse, little is known about how HIV-1 infection affects NAcc. We
Bruno A Cisterna et al.
Nature communications, 11(1), 1073-1073 (2020-02-28)
Denervation of skeletal muscles induces severe muscle atrophy, which is preceded by cellular alterations such as increased plasma membrane permeability, reduced resting membrane potential and accelerated protein catabolism. The factors that induce these changes remain unknown. Conversely, functional recovery following
X Chen et al.
Biochemistry, 33(46), 13769-13774 (1994-11-22)
Although the predominance of plasmalogens in electrically-active membranes (e.g., sarcolemma) is well-known, identification of the molecular mechanisms through which the vinyl ether linkage facilitates electrophysiologic function has remained elusive. Herein we demonstrate that the kinetics of both carrier-mediated (i.e., valinomycin)
A Minta et al.
The Journal of biological chemistry, 264(32), 19449-19457 (1989-11-15)
Fluorescent indicators sensitive to cytosolic concentrations of free Na+ have been synthesized and characterized. They consist of a crown ether, 1,7-diaza-4,10,13-trioxacyclopentadecane, linked via its nitrogens to fluorophores bearing additional liganding centers. In the currently preferred dye, SBFI (short for sodium-binding
Sung-Kyun Ko et al.
Nature chemistry, 6(10), 885-892 (2014-09-23)
Anion transporters based on small molecules have received attention as therapeutic agents because of their potential to disrupt cellular ion homeostasis. However, a direct correlation between a change in cellular chloride anion concentration and cytotoxicity has not been established for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service