Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

S0808060

Sodium stearyl maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sodium Salt of Stearyl Maleate

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C22H39NaO4
Molecular Weight:
390.53
UNSPSC Code:
41116107
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sodium stearyl maleate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium stearyl maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Pack Size

    1 answer
    1. This is a product of the European Pharmacopoeia. Each vial contains 15 mg of Sodium Stearyl Maleate.

      Please see the link below to visit the European Pharmacopeia website for more information and to download the product leaflet: https://crs.edqm.eu/db/4DCGI/View=S0808060

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service