Skip to Content
MilliporeSigma
All Photos(1)

Documents

R1148

Supelco

Rogosa SL agar

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

100
200

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.4±0.2 (25 °C)

application(s)

bioburden testing
clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

selective by low pH for Lactobacillus spp.

Application

Used as a selective medium for cultivation of Lactobacilli.

Components

Ingredients (g/L)
Tryptose, 10.00
Yeast extract, 5.00
Dextrose, 10.00
Arabinose, 5.00
Saccharose, 5.00
Sodium acetate, 15.00
Ammonium citrate, 2.00
Monopotassium phoshate, 6.00
Magnesium sulphate, 0.57
Manganese sulphate, 0.12
Ferrous sulphate, 0.03
Polysorbate 80, 1.00
Agar, 15.00

Preparation Note

Suspend 75 g of Rogosa SL Agar in 1000 ml of distilled water. Boil to dissolve the medium completely. Add 1.32 ml of glacial acetic acid (Cat. No. 45726). Mix thoroughly, and distribute into culture tubes or flasks. Heat to 90-100°C for 2-3 minutes. Cool to 45°C for direct inoculation.
DO NOT AUTOCLAVE.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A selective medium for the isolation and enumeration of oral and fecal lactobacilli.
M ROGOSA et al.
Journal of bacteriology, 62(1), 132-133 (1951-07-01)

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service