Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1127

Supelco

pH 7 Buffer

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-027-7-0.100

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

These products are discontinued but the list below shows products that can be used as replacement.

Obsolete Part NumberRelacement Part Number
QC1127-100ML1990020001
QC1127-250ML1094070500
CQ1127-500ML1094070500
CQ1127-1L1094071000
CQ1127-10L1094399010

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service