Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

QC1033

Supelco

PCBs in Water - WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 2 mL

manufacturer/tradename

RTC QCO-020

concentration

100% in acetone

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aroclor 1248

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ylenia Carotenuto et al.
Marine environmental research, 155, 104878-104878 (2020-01-25)
The sensitivity of the copepods Acartia tonsa, commonly used in standardized tests for environmental risk assessment and A. clausi, the dominant autochthonous congener species in the Mediterranean Sea, was assessed using sediment-derived elutriates from the industrial area of Bagnoli-Coroglio and
Krystyna Kozioł et al.
The Science of the total environment, 596-597, 158-168 (2017-04-23)
Glaciers accumulate organic pollutants delivered by snow. However, our understanding of the exact dynamics of organic pollutants in the snowpack relies primarily on laboratory experiments and mathematical models. To fill the gap related to the detailed field data, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service