Skip to Content
MilliporeSigma
All Photos(3)

Documents

PPB012

Sigma-Aldrich

1.0% Agarose in Tris-Acetate EDTA Buffer

pHast Pack, powder

Synonym(s):

1% Agarose gel, TAE Agarose blend

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105317
NACRES:
NA.28

product line

pHast Pack

Quality Level

form

powder

color

white to off-white

pH

8.1-8.5(range for the Tris Acetate ETDA buffer prior to blending with agarose)

solubility

soluble, cloudy to hazy, colorless to light yellow

cation traces

Fe: <10 ppm

storage temp.

room temp

General description

1% Agarose in Tris Acetate EDTA (TAE buffer, pH 8.3) is a powder blend that readily dissolves in boiling DI or ultrapure water to quickly make an agarose gel for electrophoresis. Ideal for separatation of DNA or RNA fragments that range from 400 – 8,000 bp. Gels with TAE buffer are typically used for electrophoresis of larger nucleic acid fragments.

Application

1% Agarose in 1X TAE buffer may be used to make a gel for:
  • Native RNA gel electrophoresis
  • DNA gel electrophoresis

Features and Benefits

  • pHast pack 1% agarose gel prepares 20 gels per box
  • No buffer prep or adjusting pH required
  • Saves time to cast fast, convenient and easy to use
  • Minimizes costs of multiple reagents or expensive precast gels
  • Biological and chemical tested to ensure quality: free of DNase, RNase, Protease, and Nickase
  • Compatible with TAE pHast pack buffer

Packaging

Foil pouches

Preparation Note

Prepares 250mL for casting several standard-size gels or one large agarose gel. A small electrophoresis apparatus maybe use 30 - 50mL, medium 100mL, and large rigs 250mL. Add nucleic acid stain before pouring the gel and for optimal results pair with pHast pack 1X TAE, pH 8.3 running buffer.

Reconstitution

Contents of one pouch, when dissolved in 250 mL of ultrapure water, will yield a 1× solution containing 40 mM Tris acetate, 1 mM EDTA, and 1.0% (w/v) agarose. Contents tested to be DNAse, RNAse, and Nickase free. Prepare in a 1L flask. Combine pHast pack contents with 250 mL ultrapure water and mix well. Microwave on high for ~1min to boil and dissolve the agarose. Use a heat resistant glove to remove the flask every 10 - 15s to mix gently by swirling the liquid in a circle at the bottom of the flask. Repeat until the agarose is fully dissolved. Cool the flask slightly with cold water while swirling every 10 – 15s, add the stain and pour the gel. Allow agarose gel to fully cool and solidify before removing combs.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service