Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR8965

Supelco

Dextroamphetamine Related Compound B

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

Phenylacetone, Amphetamine EP Impurity B, Benzyl methyl ketone, Phenyl-2-propanone

Sign Into View Organizational & Contract Pricing

Select a Size

PHR8965-50MG
$857.00

$857.00


Please contact Customer Service for Availability


Select a Size

Change View
PHR8965-50MG
$857.00

About This Item

Linear Formula:
C6H5CH2COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein:
742120
MDL number:
UNSPSC Code:
12352200

$857.00


Please contact Customer Service for Availability

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

(Not traceable to any primary pharmacopeia standard)

API family

dextroamphetamine, amphetamine

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

density

1.003 g/mL at 20 °C (lit.)

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

CC(=O)Cc1ccccc1

InChI

1S/C9H10O/c1-8(10)7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QCCDLTOVEPVEJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service