Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR8782

Supelco

Dipyridamole Related Compound C

new

certified reference material, pharmaceutical secondary standard

Synonym(s):

2,2′-[(6-Chloro-4,8-di-1-piperidinylpyrimido[5,4-d]pyrimidin-2-yl)imino]bisethanol, 2,2′-{[6-Chloro-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-2-yl]azanediyl}diethanol, 2-Diethanolamino-6-chloro-4,8-dipiperidinopyrimido[5,4-d]pyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30ClN7O2
CAS Number:
Molecular Weight:
435.95

Quality Level

application(s)

pharmaceutical

InChI

1S/C20H30ClN7O2/c21-19-22-15-16(17(24-19)26-7-3-1-4-8-26)23-20(28(11-13-29)12-14-30)25-18(15)27-9-5-2-6-10-27/h29-30H,1-14H2

InChI key

CWZFYGCEBHRUNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service