Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR3531

Supelco

Behenic Acid

pharmaceutical secondary standard, certified reference material

Synonym(s):

Behenic Acid, 1-Docosanoic acid, Docosanoic acid, n-Docosanoate, n-Docosanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H44O2
CAS Number:
Molecular Weight:
340.58
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 5 g

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

SMILES string

OC(=O)CCCCCCCCCCCCCCCCCCCCC

InChI key

UKMSUNONTOPOIO-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service