Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR3021

Supelco

Latanoprost Related Compound E

new

pharmaceutical secondary standard, reference material

Synonym(s):

Latanoprost acid, (5Z,9α,11α,15R)-9,11,15-Trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oic acid, (Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoic acid, PhXA 85

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
MDL number:

grade

pharmaceutical secondary standard
reference material

Quality Level

Agency

traceable to USP 1357533

API family

latanoprost

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

O[C@H](CC[C@H]1[C@H](O)C[C@H](O)[C@@H]1C\C=C/CCCC(O)=O)CCc2ccccc2

InChI

1S/C23H34O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,18-22,24-26H,2,7,10-16H2,(H,27,28)/b6-1-/t18-,19+,20+,21-,22+/m0/s1

InChI key

HNPFPERDNWXAGS-NFVOFSAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Biochem/physiol Actions

Potent, selective FP prostanoid receptor agonist, F-series prostaglandin analog. 200 times as potent as isopropyl ester form.

Packaging

Store tightly sealed, under desiccant.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service