Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1703

Supelco

Natamycin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Pimaricin, Natamycin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$269.00

$269.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$269.00

About This Item

Empirical Formula (Hill Notation):
C33H47NO13
CAS Number:
Molecular Weight:
665.73
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$269.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1457505

API family

natamycin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

C[C@@H]1C\C=C\C=C\C=C\C=C\[C@@H](C[C@@H]2O[C@](O)(C[C@@H](O)C[C@H]3O[C@@H]3\C=C\C(=O)O1)C[C@H](O)[C@H]2C(O)=O)O[C@@H]4O[C@H](C)[C@@H](O)[C@H](N)[C@@H]4O

InChI

1S/C33H47NO13/c1-18-10-8-6-4-3-5-7-9-11-21(45-32-30(39)28(34)29(38)19(2)44-32)15-25-27(31(40)41)22(36)17-33(42,47-25)16-20(35)14-24-23(46-24)12-13-26(37)43-18/h3-9,11-13,18-25,27-30,32,35-36,38-39,42H,10,14-17,34H2,1-2H3,(H,40,41)/b4-3+,7-5+,8-6+,11-9+,13-12+/t18-,19-,20+,21+,22+,23-,24-,25+,27-,28+,29-,30+,32+,33-/m1/s1

InChI key

NCXMLFZGDNKEPB-FFPOYIOWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Natamycin is a major drug that is utilized in the treatment of fungal keratitis. It is, by far, the only available approved antifungal drug for the said purpose by the U.S. Food and Drug Administration.[1]

Application

Natamycin may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high-performance liquid chromatography coupled with tandem mass spectrometry (LC−MS/MS).[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

An antifungal polyene macrolide that binds specifically to ergosterol and blocks fungal growth. However, unlike nysatin and filipin, pimaricin does not change the permeability of the plasma membrane

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2803 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioanalytical method development and validation of natamycin in rabbit tears and its application to ocular pharmacokinetic studies.
Bhatta RS, et al.
Journal of Pharmaceutical and Biomedical Analysis, 54(5), 1096-1100 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service