Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1536

Supelco

Norphenylephrine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Norphenylephrine hydrochloride, α-Aminomethyl-3-hydroxybenzyl alcohol hydrochloride, 3,α-Dihydroxyphenethylamine hydrochloride, Norfenefrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NH2)OH · HCl
CAS Number:
Molecular Weight:
189.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1473206

API family

norphenylephrine

CofA

current certificate can be downloaded

packaging

ampule of 1 × 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

162-164 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.NCC(O)c1cccc(O)c1

InChI

1S/C8H11NO2.ClH/c9-5-8(11)6-2-1-3-7(10)4-6;/h1-4,8,10-11H,5,9H2;1H

InChI key

OWMFSWZUAWKDRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Norphenylephrine Hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3654 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phenylephrine Hydrochloride
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 40(2), 3490-3490 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service