Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1386

Supelco

Amylmetacresol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Methyl-2-pentylphenol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001244

API family

amylmetacresol

CofA

current certificate can be downloaded

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

General description

Amylmetacresol is a phenolic antiseptic.[1] It is a local symptomatic pain relief throat lozenges effective in managing acute sore throat symptoms.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Amylmetacresol may be used a reference standard for the determination of amylmetacresol in bulk and pharmaceutical formulations by reverse phase-high performance liquid chromatographic (RP-HPLC) method.[1]

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3643 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Liquid Chromatographic Method for the Simultaneous Estimation of Dextromethorphan and Amylmetacresol.
Jayasree M G, et al.
International Journal of ChemTech Research, 11(01), 141-147 null
Effectiveness of amylmetacresol and 2, 4-dichlorobenzyl alcohol throat lozenges in patients with acute sore throat due to upper respiratory tract infection: a systematic review protocol.
Tan T W, et al.
JBI database of systematic reviews and implementation reports, 15(4), 862-872 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service