Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL82261

Miliacin

phyproof® Reference Substance

Synonym(s):

(3β)-3-Methoxyolean-18-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O
CAS Number:
Molecular Weight:
440.74
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (GC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

InChI

1S/C31H54O/c1-26(2)16-17-28(5)18-19-30(7)21(22(28)20-26)10-11-24-29(6)14-13-25(32-9)27(3,4)23(29)12-15-31(24,30)8/h21-25H,10-20H2,1-9H3/t21-,22?,23?,24-,25+,28-,29+,30-,31-/m1/s1

InChI key

DJAQOVIJPZOTCG-LWFXAFMDSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Molecular Evidence in Archaeological Findings: The first molecular and isotopic evidence of millet processing in prehistoric pottery vessels, possibly including traces of miliacin, provides insights into ancient agricultural practices and the dietary importance of millet in historical populations (Heron et al., 2016).

  • Miliacin′s Protective Effects in Infection: Research explores the mechanisms of the protective effect of miliacin during experimental Salmonella infection, focusing on its impact on endotoxinemia and cytokine production, valuable for understanding its potential therapeutic applications in infectious diseases (Frolov et al., 2014).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service