Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL80201

Coniine hydrochloride

phyproof® Reference Substance

Synonym(s):

(±)-2-Propylpiperidine hydrochloride, DL-Coniin hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$658.00

$658.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$658.00

About This Item

Empirical Formula (Hill Notation):
C8H17N · HCl
CAS Number:
Molecular Weight:
163.69
Beilstein:
6116710
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$658.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (GC)

manufacturer/tradename

PhytoLab

mp

~210 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CCCC1CCCCN1

InChI

1S/C8H17N.ClH/c1-2-5-8-6-3-4-7-9-8;/h8-9H,2-7H2,1H3;1H

InChI key

JXBWZNQZRWZJIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service