Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4125

Sigma-Aldrich

Patent Blue Violet (Patent Blue)

practical grade

Synonym(s):

Ethanaminium, N-[4-[[4-(diethylamino)phenyl](2,5-disulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-, inner salt, sodium salt lymphazurin, Isosulfan Blue

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C27H31N2O6S2Na
CAS Number:
Molecular Weight:
566.66
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

grade

practical grade

composition

Dye content, ~50%

mp

260 °C (dec.)

SMILES string

[Na+].CCN(CC)c1ccc(cc1)\C(=C2/C=CC(\C=C2)=[N+](\CC)CC)c3ccc(cc3S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Literature for Patent Blue Violet discusses two dyes which are very similar, Color Index Numbers 42045 and 42051. Structurally they differ with regard to the substituents on one of the aromatic rings. C.I. 42045 is 2,4-disulfo-substituted while C.I. 42051 is 2,4-disulfo-5-hydroxy-substituted.
Our NMR studies show that P 1888 and P 4125 are neither, but are instead 2,5-disulfo-substituted dyes. As yet no C.I. No. or common name has been established for this dye. We will continue to list P 1888 and P 4125 as Patent Blue Violet until the distinction has been clarified.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service