Skip to Content
MilliporeSigma
All Photos(1)

Documents

P2423

Supelco

Sigma Pseudo Narcotic Scent Cocaine formulation

powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

form

powder

Quality Level

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of cocaine.

Legal Information

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service