Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P2100000

Piperazine hydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Piperazine hexahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$205.00

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$205.00

About This Item

Empirical Formula (Hill Notation):
C4H10N2 · 6H2O
CAS Number:
Molecular Weight:
194.23
Beilstein:
4455527
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$205.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

vapor pressure

0.8 mmHg ( 20 °C)

API family

piperazine

manufacturer/tradename

EDQM

bp

145-156 °C (lit.)

mp

42-44 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].C1CNCCN1

InChI

1S/C4H10N2.6H2O/c1-2-6-4-3-5-1;;;;;;/h5-6H,1-4H2;6*1H2

InChI key

AVRVZRUEXIEGMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piperazine hydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher D Smith et al.
Clinical medicine (London, England), 14(4), 409-415 (2014-08-08)
The use of novel psychoactive substances ('legal highs' or 'designer drugs') is increasing worldwide. Patients misusing such substances have been reported to experience severe or prolonged side effects requiring admission to acute or critical care wards. These complications can be
Rajangam Vinodh et al.
Journal of nanoscience and nanotechnology, 14(6), 4639-4648 (2014-04-18)
Chloro-functionalized mesoporous MCM-41, SBA-15, MCM-48 and KIT-6 were synthesized by co-condensation of 3-chloropropyl-trimethoxy-silane (CPTMS) and rice husk ash sodium silicate solution, which is subsequently grafted with a heterocyclic amine, homopiperazine (HPZ). X-ray powder diffraction and BET analysis of the chloro-functionalized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service