Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P18608

Sigma-Aldrich

4-Phenylazomaleinanil

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N3O2
CAS Number:
Molecular Weight:
277.28
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

97%

form

crystals or chunks

mp

162-165 °C (lit.)

solubility

acetone: 25 mg/mL, clear, orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C1C=CC(=O)N1c2ccc(cc2)N=Nc3ccccc3

InChI

1S/C16H11N3O2/c20-15-10-11-16(21)19(15)14-8-6-13(7-9-14)18-17-12-4-2-1-3-5-12/h1-11H

InChI key

DVNPYLMPVFDKGZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M O'Malley et al.
Biochemical and biophysical research communications, 200(1), 384-388 (1994-04-15)
A nonfluorescent reagent, 4-phenylazophenylmaleimide [4-PAPM], was attached to the sole cysteine residue [104(G11)] of alpha apohemoglobin (alpha degree) and served as an energy acceptor for the single intrinsic tryptophanyl [14(A12)] donor. This novel fluorescence system provided a transmolecular vehicle by
Masafumi D Yamada et al.
Journal of biochemistry, 142(6), 691-698 (2007-10-19)
Azobenzene is a photochromic molecule that undergoes rapid and reversible isomerization between the cis- and trans-forms in response to ultraviolet (UV) and visible (VIS) light irradiation, respectively. Here, we introduced the sulfhydryl-reactive azobenzene derivative 4-phenylazophenyl maleimide (PAM) into the functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service