Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P1347

Sigma-Aldrich

Potassium bicarbonate

meets USP testing specifications

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352300

Agency

meets USP testing specifications

application(s)

pharmaceutical (small molecule)

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Ranjit et al.
Vascular pharmacology, 71, 102-107 (2015-04-15)
Research and routine laboratory assessment of clot integrity can be time consuming, expensive, and cannot be batched as it is generally performed in real time. To address these issues, we developed and validated a micro-titre based assay to quantify thrombogenesis
Hadi Shafiee et al.
Scientific reports, 5, 9919-9919 (2015-06-06)
We report a biosensing platform for viral load measurement through electrical sensing of viruses on a flexible plastic microchip with printed electrodes. Point-of-care (POC) viral load measurement is of paramount importance with significant impact on a broad range of applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service