Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NISTRM8188

Soy-containing solid oral dosage form

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 13 mg

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Reference/Informational value is provided for the following analytes.
Daidzein (C15H10O4), Daidzin(a) (C21H20O9), Genistein (C15H10O5), Genistin(a) (C21H20O10), Glycitein (C16H12O5), Glycitin (C22H22O10)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service