NISTC2418
High-purity lead
NIST® SRM® C2418
Synonym(s):
Lead
About This Item
Recommended Products
grade
certified reference material
packaging
pkg of disk
manufacturer/tradename
NIST®
resistivity
20.65 μΩ-cm
technique(s)
optical emission spectroscopy (OES): suitable
bp
1740 °C (lit.)
mp
327.4 °C (lit.)
format
matrix material
SMILES string
[Pb]
InChI
1S/Pb
InChI key
WABPQHHGFIMREM-UHFFFAOYSA-N
Related Categories
General description
SRM C2418_cert
SRM C2418_SDS
Other Notes
Cadmium (Cd), silver (Ag)
Reference/Informational value is provided for the following analytes.
Aluminium (Al), Antimony (Sb), Arsenic (As), Bismuth (Bi), Calcium (Ca), Cobalt (Co), Copper (Cu), Iron (Fe), Manganese (Mn), Nickel (Ni), Tellurium (Te), Tin (Sn), Zinc (Zn)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral
Target Organs
Central nervous system,Blood,Immune system,Kidney
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service