Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST977

Isotopic standard for bromine

NIST® SRM® 977

Synonym(s):

Sodium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

0.25 G
$987.00

$987.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.25 G
$987.00

About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.24

$987.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

vapor pressure

1 mmHg ( 806 °C)

isotopic purity

 79Br
 81Br

packaging

pkg of 0.25 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

mp

755 °C (lit.)

format

matrix material

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Bromine-79 (79Br), Bromine-81 (81Br)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service