Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST92

Soda-lime glass

NIST® SRM® 92, (low boron)

Sign Into View Organizational & Contract Pricing

Select a Size

45 G
$1,390.00

$1,390.00


Estimated to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
45 G
$1,390.00

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

$1,390.00


Estimated to ship onApril 23, 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 45 g

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

glass & ceramic

format

matrix material

storage temp.

15-25°C

SMILES string

[Ca+2].[Na+].[P](=O)([O-])([O-])[O-]

InChI

1S/Ca.Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q+2;+1;/p-3

InChI key

QXJJQWWVWRCVQT-UHFFFAOYSA-K

General description

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Arsenic (As), Calcium (Ca), Iron (Fe), Lead (Pb), Magnesium (Mg), Potassium (K), Silicon (Si), Sodium (Na), Sulfur (S), Boron oxide (B2O3)

Reference/Informational value is provided for the following analytes.
Barium (Ba), Manganese (Mn), Phosphorus (P), Strontium (Sr), Titanium (Ti), Zinc (Zn), Zirconium (Zr), Antimony (Sb), Gallium (Ga), Beryllium (Be), Fluorine (F), Bismuth (Bi), Lithium (Li), Cadmium (Cd), Mercury (Hg), Cerium (Ce), Nickel (Ni), Cesium (Cs), Rubidium (Rb), Chromium (Cr), Selenium (Se), Cobalt (Co), Tin (Sn), Copper (Cu), Vanadium (V)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service