Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST8495

Northern Softwood Bleached Kraft Pulp

NIST®SRM®

Synonym(s):

Cellulose pulp

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 5 kg (10 sheets)

manufacturer/tradename

NIST®

sheet thickness

0.937 mm

density

718 kg/m3

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Northern Softwood Bleached Kraft Pulp from NIST. Safety Information available. See Certificate/Report for additional Information. Excerpt below: These Reference Materials (RMs) are intended primarily for use in fundamental studies on the properties of fibers and paper sheets. The RMs provide samples so that investigators in different laboratories can be assured that they are investigating the same materials. The materials selected for these two RMs are bleached dried lap pulp each from a single lot of a standard commercial production run. Each RM package consists of ten standard lap sheets. Each sheet weighs approximately one pound bone dry. The ten sheets are hermetically-sealed in a film foil barrier bag and packaged in a 200 pound C flute carton with dust flaps. The carton is \H\ taped to eliminate dust and light penetration. Each carton of ten sheets weighs approximately five kilograms. The materials were selected for the two RMs because of their differing fiber size, differing papermaking properties, and similarity to commercially available materials.

SRM 8495_cert

SRM 8495_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service