Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST8301

Boron Isotopes in Marine Carbonate (Simulated Coral and Foraminifera Solutions)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

24 G
$1,160.00

$1,160.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
24 G
$1,160.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,160.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

isotopic purity

 11B

packaging

pkg of 24 g

manufacturer/tradename

NIST®

format

multi-component solution

storage temp.

15-25°C

General description

Boron Isotopes in Marine Carbonate (Simulated Coral and Foraminifera Solutions) The Standard Reference Material (SRM) is intended for use in developing and evaluating methods to measure relative differences in boron (B) isotope-number ratios, specifically R(¹¹B/¹⁰B). For more information, please refer to the SDS and COA.

SRM 8301_cert

SRM 8301_SDS

Application

Each unit of RM 8301 includes three vials, each containing two simulated solutions labeled RM 8301 (Coral) and RM 8301 (Foram). Each vial holds approximately 4 mL of a gravimetrically prepared solution with a nominal mass fraction of 50 mg/g of calcium, dissolved in approximately 3 mol/L nitric acid in water.

Features and Benefits

  • Reference values and expanded uncertainties for δ11BSRM951 are determined using the DerSimonian-Laird analysis method within the NIST Consensus Builder.
  • Expiration details, instructions for handling, storage and use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Boron (11B/10B)

Reference/Informational value is provided for the following analytes.
Boron (B), Calcium (Ca)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service