Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST69B

Bauxite

NIST® SRM® 69b, Arkansas

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of fine powder (nominal particle size < 0.08 mm) and is intended for use in evaluating chemical and instrumental methods of analyses. For more information, please refer to the SDS and C

SRM 69B_cert

SRM 69B_SDS

Other Notes

Certified for the analytes listed below.
Aluminium oxide (Al2O3), Ferric oxide (Fe2O3), Silicon dioxide (SiO2), Titanium dioxide (TiO2), Zirconium Dioxide (ZrO2), Phosphorus pentoxide (P2O5), Vanadium pentoxide (V2O5), Chromium oxide (Cr2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Manganese oxide (MnO), Zinc oxide (ZnO), Potassium oxide (K2O), Sulfur trioxide (SO3)

Reference/Informational value is provided for the following analytes.

Barium oxide (BaO), Sodium oxide (Na2O), Cerium (Ce), Cobalt (Co), Hafnium (Hf), Scandium (Sc)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service